Anti-TBX3 Mouse Monoclonal Antibody [clone: 7H6]
ABNOH00006926-M03
: Abnova
- Antibody type:Primary
- Antigen name:T-box 3
- Antigen symbol:TBX3
- Clonality:Monoclonal
- Clone:7H6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6926
- Antigen synonyms:XHL|TBX3-ISO|UMS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TBX3
Clonality: Monoclonal
Clone: 7H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human