Anti-CK33B Mouse Monoclonal Antibody [clone: 4A6-A7]
Catalog # ABNOH00003884-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 33B
- Antigen symbol:CK33B
- Clonality:Monoclonal
- Clone:4A6-A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3884
- Antigen synonyms:K33B|hHa3-II|Ha-3II|HA3II|KRTHA3A|KRTHA3B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CK33B
Clonality: Monoclonal
Clone: 4A6-A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human