Anti-IL15 Mouse Monoclonal Antibody [clone: 3B9]
Catalog # ABNOH00003600-M12
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 15
- Antigen symbol:IL15
- Clonality:Monoclonal
- Clone:3B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:3600
- Antigen synonyms:IL-15
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IL15
Clonality: Monoclonal
Clone: 3B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human