Anti-ID1 Mouse Monoclonal Antibody [clone: 2C7]
ABNOH00003397-M11
: Abnova
- Antibody type:Primary
- Antigen name:Inhibitor Of Differentiation 1
- Antigen symbol:ID1
- Clonality:Monoclonal
- Clone:2C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3397
- Antigen synonyms:BHLHb24|Dominant Negative Helix-Loop-Helix Protein|Class B Basic Helix-Loop-Helix Protein 24|DNA-Binding Protein Inhibitor ID-1|Inhibitor Of DNA Binding 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ID1
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human