Order Entry
Export
ContactUsLinkComponent
Anti-GFRA1 Mouse Monoclonal Antibody [clone: 3D8]
Anti-GFRA1 Mouse Monoclonal Antibody [clone: 3D8]
Catalog # ABNOH00002674-M11
Supplier:  Abnova
undefined
Anti-GFRA1 Mouse Monoclonal Antibody [clone: 3D8]
Catalog # ABNOH00002674-M11
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GDNF family receptor alpha 1
  • Antigen symbol:
    GFRA1
  • Clonality:
    Monoclonal
  • Clone:
    3D8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    2674
  • Antigen synonyms:
    GFR-ALPHA-1|RETL1|GDNFR|RET1L|GDNFRA|TRNR1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: GFRA1
Clonality: Monoclonal
Clone: 3D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human