Anti-FH Mouse Monoclonal Antibody [clone: 2E10]
Catalog # ABNOH00002271-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fumarate hydratase
- Antigen symbol:FH
- Clonality:Monoclonal
- Clone:2E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2271
- Antigen synonyms:LRCC|FMRD|MCL|HLRCC|MCUL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FH
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human