Anti-TRIM44 Mouse Monoclonal Antibody [clone: OTI2B9]
ORIGTA505513
: OriGene
- Antibody type:Primary
- Antigen name:tripartite motif containing 44
- Antigen symbol:TRIM44
- Clonality:Monoclonal
- Clone:OTI2B9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:38.3 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Type: Control
Antigen: TMX1 (thioredoxin-related transmembrane protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: TRIM44 (tripartite motif containing 44)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human