Anti-SETD7 Mouse Monoclonal Antibody [clone: OTI2D10]
Catalog # ORIGTA503322
Supplier: OriGene
Jump to:
Product Details & Documents
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SETD7 (SET domain containing (lysine methyltransferase) 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Documents
TA503322.PDF
Datasheet
Specifications
- Western blot:Yes
- Clonality:Monoclonal
- Flow cytometry:Yes
- Purification:purified by affinity chromatography
- Antibody type:Primary
- Epitope:Full length
- Form:Liquid
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:none
- Molecular weight:40.5 kDa
- Shipping temperature:Ice
- Antigen name:SET domain containing (lysine methyltransferase) 7
- Cat. No.:ORIGTA503322
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:SETD7
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Clone:OTI2D10



