Anti-IFT57 Mouse Monoclonal Antibody [clone: OTI4F6]
Catalog # ORIGTA500918
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:intraflagellar transport 57 homolog (Chlamydomonas)
- Antigen symbol:IFT57
- Clonality:Monoclonal
- Clone:OTI4F6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:49.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: IFT57 (intraflagellar transport 57 homolog (Chlamydomonas))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human