Order Entry
Export
ContactUsLinkComponent
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI2C8]
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI2C8]
  ORIGTA500457
 :  OriGene
undefined
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI2C8]
  ORIGTA500457
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    nuclear receptor binding protein 1
  • Antigen symbol:
    NRBP1
  • Clonality:
    Monoclonal
  • Clone:
    OTI2C8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    59.8 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1 (nuclear receptor binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human