Anti-NEUROG1 Mouse Monoclonal Antibody [clone: OTI6C9]
Catalog # ORIGTA500306
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Neurogenin 1
- Antigen symbol:NEUROG1
- Clonality:Monoclonal
- Clone:OTI6C9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:25.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg
Type: Primary
Antigen: NEUROG1 (neurogenin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human