Anti-UBC Rabbit Polyclonal Antibody
Catalog # ORIGTA317483
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin C
- Antigen symbol:UBC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:near the C-terminus
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Pk:50 µG
Specifications
About this item
Recommended Dilutions: Immunochemistry: 10 µg/ml; Western Blot: 1:100-1:500
Type: Primary
Antigen: UBC (ubiquitin C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human