Anti-DICER1 Rabbit Polyclonal Antibody
Catalog # ORIGTA310112
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:dicer 1, ribonuclease type III
- Antigen symbol:DICER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, pH 7,4 with 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:100-1:500
Type: Primary
Antigen: DICER1 (dicer 1, ribonuclease type III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse