Order Entry
Export
ContactUsLinkComponent
Anti-SHANK3 Mouse Monoclonal Antibody [clone: S69-46]
Anti-SHANK3 Mouse Monoclonal Antibody [clone: S69-46]
Catalog # ORIGTA309476
Supplier:  OriGene
Anti-SHANK3 Mouse Monoclonal Antibody [clone: S69-46]
Catalog # ORIGTA309476
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SH3 and multiple ankyrin repeat domains 3
  • Antigen symbol:
    SHANK3
  • Clonality:
    Monoclonal
  • Clone:
    S69-46
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    840 to 857
  • Storage buffer:
    PBS pH 7,4, 50% glycerol, 0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions:

Type: Primary
Antigen: SHANK3 (SH3 and multiple ankyrin repeat domains 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: