Anti-NR2B Mouse Monoclonal Antibody [clone: S59-36]
Catalog # ORIGTA309448
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:NR2B
- Clonality:Monoclonal
- Clone:S59-36
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: NR2B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: