Order Entry
Export
ContactUsLinkComponent
Anti-NR2B Mouse Monoclonal Antibody [clone: S59-20]
Anti-NR2B Mouse Monoclonal Antibody [clone: S59-20]
  ORIGTA309447
 :  OriGene
Anti-NR2B Mouse Monoclonal Antibody [clone: S59-20]
  ORIGTA309447
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    NR2B
  • Clonality:
    Monoclonal
  • Clone:
    S59-20
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS pH 7,4, 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Recommended Dilutions:

Type: Primary
Antigen: NR2B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat