Anti-CNGA1 Mouse Monoclonal Antibody [clone: S36-12]
Catalog # ORIGTA309315
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:cyclic nucleotide gated channel alpha 1
- Antigen symbol:CNGA1
- Clonality:Monoclonal
- Clone:S36-12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: CNGA1 (cyclic nucleotide gated channel alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: