Anti-CACNA1C Mouse Monoclonal Antibody [clone: S57-46]
ORIGTA309306
: OriGene
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, L type, alpha 1C subunit
- Antigen symbol:CACNA1C
- Clonality:Monoclonal
- Clone:S57-46
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml
Type: Primary
Antigen: CACNA1C (calcium channel, voltage-dependent, L type, alpha 1C subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat