Anti-PODXL Mouse Monoclonal Antibody [clone: TRA-1-60(R)]
Catalog # ORIGTA301484
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Podocalyxin-like 1
- Antigen symbol:PODXL1
- Clonality:Monoclonal
- Clone:TRA-1-60(R)
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Storage buffer:Tris-glycine, 150 mM NaCl, 0,05% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: PODXL (podocalyxin-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human