Order Entry
Export
ContactUsLinkComponent
 
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4]
  70-0048-U100
undefined
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4]
  70-0048-U100

 

  • Antibody type:
    Primary
  • Antigen name:
    CD4 molecule
  • Antigen symbol:
    CD4
  • Clonality:
    Monoclonal
  • Clone:
    OKT4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Cynomolgus monkey,
    Rhesus monkey,
    Chimpanzee,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    920
  • Antigen synonyms:
    L3T4|p55|CD4mut|Ly-4|W3/25
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

The OKT4 antibody reacts with human CD4, a 59 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4, and co-receptors CCR5 and CXCR4, may also be utilized by HIV-1 to enter T cells. Human CD4 is typically expressed on thymocytes, some mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.

The OKT4 antibody is widely used as a phenotypic marker for CD4 expression. It is cross-reactive with CD4 in several non-human species, including Chimpanzee, Cynomolgus and Rhesus. This antibody recognizes a different epitope, and thus does not block binding of, the alternative Anti-Human CD4 antibody clone RPA-T4 (Reinherz EL, et al. 1979. Proc. Natl. Acad. Sci. 76:4061-4065).

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: OKT4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: