Order Entry
Export
ContactUsLinkComponent
Anti-CD28 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CD28.2]
Anti-CD28 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CD28.2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD28 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CD28.2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0289-T025
  • 20-0289-T100
  • Antigen symbol:
  • CD28
  • CD28
  • Antigen name:
  • CD28 Molecule
  • CD28 Molecule
  • Antigen synonyms:
  • Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
  • Tp44|MGC138290|T-cell-specific surface glycoprotein CD28|CD28 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • CD28.2
  • CD28.2
  • Reactivity:
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 940
  • 940
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Purification:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Cool
  • Cool
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information