Order Entry
Export
ContactUsLinkComponent
Anti-K0776 Rabbit Polyclonal Antibody
Anti-K0776 Rabbit Polyclonal Antibody
  ABGEAP9677C
 :  Abgent
undefined
Anti-K0776 Rabbit Polyclonal Antibody
  ABGEAP9677C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Nonhomologous End-Joining Factor 1
  • Antigen symbol:
    KIAA0776
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Gene ID:
    79840
  • Antigen synonyms:
    XLF
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    RPQLSKVKRKKPRGC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This NHEJ1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 268-296 amino acids from the C-terminal region of human NHEJ1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: K0776
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human