Anti-GPR133 Rabbit Polyclonal Antibody
ABGEAP9499C
: Abgent
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 133
- Antigen symbol:GPR133
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:23327
- Antigen synonyms:GPR133|adhesion G protein-coupled receptor D1|PGR25
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:VPYSREFKQKYDYFC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NEDD4L antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 576-604 amino acids from the Central region of human NEDD4L.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: GPR133 (G protein-coupled receptor 133)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human