Anti-CDC20 Rabbit Polyclonal Antibody
ABGEAP9138A
: Abgent
- Antibody type:Primary
- Antigen name:Cell division cycle 20
- Antigen symbol:CDC20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:75770
- Antigen synonyms:CDC20A|p55CDC|bA276H19.3
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:RYPSHEDEDLPPRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Brsk2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 329-356 amino acids from the Central region of mouse Brsk2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: CDC20 (cell division cycle 20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human