Anti-C1QTNF6 Rabbit Polyclonal Antibody
Catalog # ABGEAP8883A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:C1q tumor necrosis factor-related protein 6
- Antigen symbol:C1QTNF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:31
- Antigen synonyms:CTRP6|CTFP6|UNQ581/PRO1151|Complement C1q tumor necrosis factor-related protein 6|ZACRP6
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:KTYQAIKDFNREGLPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ACACA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 2010-2039 amino acids from the C-terminal region of human ACACA.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: C1QTNF6 (C1q and tumor necrosis factor related protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human