Anti-DGKD Rabbit Polyclonal Antibody
ABGEAP8126B
: Abgent
- Antibody type:Primary
- Antigen name:Diacylglycerol kinase, delta 130kDa
- Antigen symbol:DGKD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal (1000–1100 aa)
- Cross adsorption:No
- Gene ID:632
- Antigen synonyms:130 kDa diacylglycerol kinase|DAG kinase delta|Diacylglycerol kinase delta|Diglyceride kinase
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:SKQEGSEVVKRPRRYC
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This BGLAP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 23-52 amino acids from the Central region of human BGLAP.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: DGKD (diacylglycerol kinase, delta 130kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse