Anti-PLK3 Rabbit Polyclonal Antibody
ABGEAP8044B
: Abgent
- Antibody type:Primary
- Antigen name:Polo-like kinase 3
- Antigen symbol:PLK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Gene ID:7764
- Antigen synonyms:FNK|CNK|Proliferation-related kinase|FGF-inducible kinase|Cytokine-inducible serine/threonine-protein kinase|PRK
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:PLDKSQVRTEPPKEC
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This ZNF217 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 92-120 amino acids from the N-terminal region of human ZNF217.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: PLK3 (polo-like kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse