Anti-RET Rabbit Polyclonal Antibody
Catalog # ABGEAP7669A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ret Proto-Oncogene
- Antigen symbol:RET
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal C166 (166–181 aa)
- Cross adsorption:No
- Antigen synonyms:MEN2B|RET51|RET-ELE1|MEN2A|CDHF12|CDHR16|PTC|MTC1|HSCR1
- Sequence:C166
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RET (ret proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human