Order Entry
Export
ContactUsLinkComponent
Anti-ILK2/ILK1 Rabbit Polyclonal Antibody
Anti-ILK2/ILK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7077B
Supplier:  Abgent
undefined
Anti-ILK2/ILK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7077B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin-Linked Kinase
  • Antigen symbol:
    ILK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Antigen synonyms:
    ILK1|Integrin-linked protein kinase|ILK2|p59ILK |59 kDa serine/threonine-protein kinase|ILK-1|ILK-2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ILK2/ILK1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse