Anti-AKT2 Rabbit Polyclonal Antibody
Catalog # ABGEAP7029C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:V-Akt Murine Thymoma Viral Oncogene Homolog 2
- Antigen symbol:AKT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center (100–200 aa)
- Cross adsorption:No
- Antigen synonyms:RAC-BETA|PKBB|PRKBB|PKBBETA|HIHGHH
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human