Anti-CR2 Rabbit Polyclonal Antibody
ABGEAP6965B
: Abgent
- Antibody type:Primary
- Antigen name:Complement Component (3d/Epstein Barr virus) receptor 2
- Antigen symbol:CR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:SLEB9|CR|CVID7|CR2|CD21|C3DR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CR2 (complement component (3d/Epstein Barr virus) receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human