Anti-KPNB1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6816A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Karyopherin beta 1
- Antigen symbol:KPNB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:Impnb|IPOB|IMB1|NTF97|IPO1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: KPNB1 (karyopherin (importin) beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human