Anti-RCBTB2 Rabbit Polyclonal Antibody
Catalog # ABGEAP6741A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:RCC1 and BTB domain-containing protein 2
- Antigen symbol:RCBTB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:ARH, ARH1, ARH2, FHCB1, FHCB2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RCBTB2 (regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human