Anti-KRT14 Rabbit Polyclonal Antibody
Catalog # ABGEAP6713C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 14
- Antigen symbol:CK14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:K14|NFJ|EBS4|CK14|EBS3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: KRT14 (keratin 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human