Anti-SMYD5 (RAI15) Rabbit Polyclonal Antibody
Catalog # ABGEAP6234A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:SET And MYND Domain Containing 5
- Antigen symbol:SMYD5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:NN8-4AG|RAI15|ZMYND23|RRG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SMYD5 (RAI15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse