Anti-CREB3L1 Rabbit Polyclonal Antibody
ABGEAP6229A
: Abgent
- Antibody type:Primary
- Antigen name:cAMP responsive element binding protein 3-like 1
- Antigen symbol:CREB3L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:Processed cyclic AMP-responsive element-binding protein 3-like protein 1|Old astrocyte specifically-induced substance|OASIS|Cyclic AMP-responsive element-binding protein 3-like protein 1|cAMP-responsive element-binding protein 3-like protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CREB3L1 (cAMP responsive element binding protein 3-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human