Anti-HAND2 Rabbit Polyclonal Antibody
Catalog # ABGEAP6117A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Heart and neural crest derivatives expressed 2
- Antigen symbol:HAND2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:Mothers against decapentaplegic homolog 2|hMAD-2|hSMAD2|JV18-1|MADR2|MADH2|MGC34440|Mothers against DPP homolog 2|MGC22139|JV18|Mad-related protein 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: HAND2 (heart and neural crest derivatives expressed 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human