Anti-SIA8C Rabbit Polyclonal Antibody
Catalog # ABGEAP5596B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Cell division cycle 42
- Antigen symbol:CDC42
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:G25K GTP-binding protein|G25K|CDC42Hs|Cell division control protein 42 homolog
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SIA8C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human