Anti-SRD5A2 Rabbit Polyclonal Antibody
ABGEAP5552C
: Abgent
- Antibody type:Primary
- Antigen name:Steroid 5 alpha-reductase 2
- Antigen symbol:SRD5A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:SRD5A2|MGC138457|alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2)|steroid-5-alpha-reductase|3-oxo-5 alpha-steroid 4-dehydrogenase 2|steroid 5-alpha-reductase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SRD5A2 (steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse