Anti-BTG1 Rabbit Polyclonal Antibody
Catalog # ABGEAP5111C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Glucosamine (N-acetyl)-6-sulfatase
- Antigen symbol:GNS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:N-acetylglucosamine-6-sulfatase|G6S|GNS|Glucosamine-6-sulfatase
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: BTG1 (B-cell translocation gene 1, anti-proliferative)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse