Anti-AVIL Rabbit Polyclonal Antibody
ABGEAP4756A
: Abgent
- Antibody type:Primary
- Antigen name:Advillin
- Antigen symbol:AVIL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:AVIL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: AVIL (advillin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human