Anti-SC24A Rabbit Polyclonal Antibody
ABGEAP4701C
: Abgent
- Antibody type:Primary
- Antigen name:SEC24 family member A
- Antigen symbol:SEC24A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:heat shock protein J2|dnaJ homolog subfamily A member 1|heat shock protein|HSJ-2|dnaJ protein homolog 2|DNAJ-like 2|neural precursor cell expressed|human DnaJ protein 2|heat shock 40 kDa protein 4|developmentally down-regulated 7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SC24A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human