Anti-RAF1 Rabbit Polyclonal Antibody
ABGEAP3445A
: Abgent
- Antibody type:Primary
- Antigen name:RAF proto-oncogene serine/threonine-protein kinase
- Antigen symbol:RAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:Raf-1|CRAF|RAF|NS5|cRaf|C-raf|c-Raf|Proto-oncogene c-RAF
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human