Order Entry
Export
ContactUsLinkComponent
Anti-RB1 Rabbit Polyclonal Antibody
Anti-RB1 Rabbit Polyclonal Antibody
  ABGEAP3241A
 :  Abgent
undefined
Anti-RB1 Rabbit Polyclonal Antibody
  ABGEAP3241A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Retinoblastoma 1
  • Antigen symbol:
    RB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    RB1|pRb|OSRC|pp110|PP110|P105-RB|RB|Retinoblastoma-associated protein|p105-Rb
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human