Order Entry
Export
ContactUsLinkComponent
Anti-EIF4E2 Rabbit Polyclonal Antibody
Anti-EIF4E2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1955A
Supplier:  Abgent
undefined
Anti-EIF4E2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1955A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ribosomal Protein S6 Kinase, 70kDa, Polypeptide 1
  • Antigen symbol:
    RPS6KB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    mRNA cap-binding protein 4EHP|EIF4E2|mRNA cap-binding protein type 3|eIF-4E type 2|eIF4E-like protein 4E-LP|EIF4EL3|Eukaryotic translation initiation factor 4E type 2|Eukaryotic translation initiation factor 4E-like 3|eIF4E type 2|Eukaryotic translation initiation factor 4E homologous protein
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: EIF4E2 (eukaryotic translation initiation factor 4E family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human