Order Entry
Export
ContactUsLinkComponent
Anti-BECN1 Rabbit Polyclonal Antibody
Anti-BECN1 Rabbit Polyclonal Antibody
Catalog # ABGEAP1818A
Supplier:  Abgent
undefined
Anti-BECN1 Rabbit Polyclonal Antibody
Catalog # ABGEAP1818A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Beclin 1
  • Antigen symbol:
    BECN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Antigen synonyms:
    Coiled-coil myosin-like BCL2-interacting protein|VPS30|ATG6|GT197|Beclin-1|beclin1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse