Order Entry
Export
ContactUsLinkComponent
Anti-ASNS Rabbit Polyclonal Antibody
Anti-ASNS Rabbit Polyclonal Antibody
Catalog # ABGEAP16927A
Supplier:  Abgent
undefined
Anti-ASNS Rabbit Polyclonal Antibody
Catalog # ABGEAP16927A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Asparagine Synthetase (Glutamine-Hydrolyzing)
  • Antigen symbol:
    ASNS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Gene ID:
    440
  • Antigen synonyms:
    ASNSD, TS11
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    64.37 kDa
  • Sequence:
    LGRDTYGVRPLFKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ASNS antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 120-147 amino acids from the N-terminal region of human ASNS.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ASNS (asparagine synthetase (glutamine-hydrolyzing))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human