Anti-HPS3 Rabbit Polyclonal Antibody
Catalog # ABGEAP16879A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Hermansky-Pudlak syndrome 3
- Antigen symbol:HPS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:84343
- Antigen synonyms:SUTAL
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:113.736 kDa
- Sequence:TGLADEKRKYSHFQC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This HPS3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 262-290 amino acids from the N-terminal region of human HPS3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: HPS3 (Hermansky-Pudlak syndrome 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human