Anti-SPRR3 Rabbit Polyclonal Antibody
ABGEAP16124B
: Abgent
- Antibody type:Primary
- Antigen name:Small Proline-Rich Protein 3
- Antigen symbol:SPRR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:6707
- Antigen synonyms:22 kDa pancornulin|Small proline rich protein 3|Small proline-rich protein 3|SPRC|Cornin beta|Cornin B|Sprr3|SPRR3_HUMAN.|Esophagin
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:18.154 kDa
- Sequence:KVPDQGFIKFPEPGC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SPRR3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 101-129 amino acids from the C-terminal region of human SPRR3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: SPRR3 (small proline-rich protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human