Anti-XPO5 Rabbit Polyclonal Antibody
Catalog # ABGEAP16050B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Exportin 5
- Antigen symbol:XPO5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:57510
- Antigen synonyms:Exp 5|Exportin 5|xpo5|Exp5|Exportin5|Xpo 5|Ran-binding protein 21|Exportin-5|Ranbp 21|FLJ14239|XPO5_HUMAN.|Ran binding protein 21|FLJ45606|Ranbp21|KIAA1291|FLJ32057
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:136.311 kDa
- Sequence:LQKVADKRRKDQFKRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This XPO5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1123-1152 amino acids from the C-terminal region of human XPO5.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: XPO5 (exportin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human